Learn More
Immunoreagents Inc GOAT A-HUMAN IGG AP 2 MG
Supplier:  Immunoreagents Inc GTXHU004F
Encompass_Preferred
Secondary Antibody; Goat anti-human IgG, Fc - Affinity Pure, min x w/human IgA + IgM, Host: Goat, Format: Liq, Label: None , Applications: WB, ELISA, IHC/ICC, IF, IP, FC
Catalog No. 50-271-119