Learn More
Immunoreagents Inc GOAT A-HUMAN IGG DYLIGHT 633
Supplier:  Immunoreagents Inc GTXHU004D633NHSX
Encompass_Preferred
Secondary Antibody; Goat anti-human IgG, Fc - Affinity Pure, DyLight 633 Conjugate, Host: Goat, Format: Lyo, Label: DyL633, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-272-055