Learn More
Immunoreagents Inc GOAT A-HUMAN IGG DYLIGHT 650
Supplier:  Immunoreagents Inc GTXHU004F2650NHSX
Encompass_Preferred
Secondary Antibody; Goat anti-human IgG, Fc - Affinity Pure, min x w/human IgA + IgM, Bo, Eq, Mu or Rb serum proteins, DyLight 650 Conjugate, Host: Goat, Format: Lyo, Label: DyL650, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-270-862