Learn More
Immunoreagents Inc GOAT A-HUMAN IGG DYLIGHT 800
Supplier:  Immunoreagents Inc GTXHU003E800NHSX
Encompass_Preferred
Secondary Antibody; Goat anti-human IgG, (H&L), Affinity Pure, min x w/Bo, Mu or Rb serum proteins, DyLight 800 Conjugate, Host: Goat, Format: Lyo, Label: DyL800, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-270-948