Learn More
Immunoreagents Inc GOAT A-HUMAN IGG FITC 0.5 MG
Supplier:  Immunoreagents Inc GTXHU003GFITC
Encompass_Preferred
Secondary Antibody; Goat anti-human IgG, (H&L), F(ab)'2 fragment - Affinity Pure, FITC Conjugate, Host: Goat, Format: Liq, Label: FITC, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-270-963