Learn More
Immunoreagents Inc GOAT A-HUMAN IGG FITC 0.5 MG
Supplier:  Immunoreagents Inc GTXHU004J2FITC
Encompass_Preferred
Secondary Antibody; Goat anti-human IgG, Fc, F(ab)'2 fragment - Affinity Pure, min x w/Bo, Eq, Mu or Rb serum proteins, Mu IgG1, FITC Conjugate, Host: Goat, Format: Liq, Label: FITC, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-271-848