Learn More
Immunoreagents Inc GOAT A-HUMAN IGG FITC 2 MG
Supplier:  Immunoreagents Inc GTXHU004EFITC
Encompass_Preferred
Secondary Antibody; Goat anti-human IgG, Fc - Affinity Pure, min x w/Bo, Eq, Mu or Rb serum proteins, FITC Conjugate, Host: Goat, Format: Liq, Label: FITC, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-271-475