Learn More
Immunoreagents Inc GOAT A-HUMAN IGM MU ALP
Supplier:  Immunoreagents Inc GTXHU006EALP
Encompass_Preferred
Secondary Antibody; Goat anti-human IgM, (muchain), Affinity Pure, min x w/Human IgA + IgG, ALP Conjugate, Host: Goat, Format: Liq, Label: ALP, Applications: WB, ELISA
Catalog No. 50-271-497