Learn More
Immunoreagents Inc GOAT A-HUMAN IGM MU AP
Catalog No. 50271850
Encompass_Preferred
Secondary Antibody; Goat anti-human IgM, (muchain), F(ab)'2 fragment - Affinity Pure, min x w/IgG from Hu or Mu, Host: Goat, Format: Liq, Label: None , Applications: WB, ELISA, IHC/ICC, IF, IP, FC
Catalog No. 50-271-850 Supplier Immunoreagents Inc Supplier No. GTXHU006J3
Secondary Antibody; Goat anti-human IgM, (muchain), F(ab)'2 fragment - Affinity Pure, min x w/IgG from Hu or Mu, Host: Goat, Format: Liq, Label: None , Applications: WB, ELISA, IHC/ICC, IF, IP, FC