Learn More
Immunoreagents Inc GOAT A-HUMAN IGM MU AP
Supplier: Immunoreagents Inc GTXHU006J
Secondary Antibody; Goat anti-human IgM, (muchain), F(ab)'2 fragment - Affinity Pure, min x w/Human IgA + IgG, Host: Goat, Format: Liq, Label: None , Applications: WB, ELISA, IHC/ICC, IF, IP, FC