Learn More
Immunoreagents Inc GOAT A-HUMAN IGM MU FITC
Supplier:  Immunoreagents Inc GTXHU006JFITC
Encompass_Preferred
Secondary Antibody; Goat anti-human IgM, (muchain), F(ab)'2 fragment - Affinity Pure, min x w/Human IgA + IgG, FITC Conjugate, Host: Goat, Format: Liq, Label: FITC, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-272-007