Learn More
Immunoreagents Inc GOAT A-HUMAN IGM MU HRP
Supplier:  Immunoreagents Inc GTXHU006JHRPX
Encompass_Preferred
Secondary Antibody; Goat anti-human IgM, (muchain), F(ab)'2 fragment - Affinity Pure, min x w/Human IgA + IgG, HRP Conjugate, Host: Goat, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-911