Learn More
Immunoreagents Inc GOAT A-HUMAN IGM MU HRP
Supplier:  Immunoreagents Inc GTXHU006DHRPX
Encompass_Preferred
Secondary Antibody; Goat anti-human IgM, (muchain), Affinity Pure, HRP Conjugate, Host: Goat, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-270-693