Learn More
Immunoreagents Inc GOAT A-MOUSE IGG ALP
Supplier:  Immunoreagents Inc GTXMU003JALP
Encompass_Preferred
Secondary Antibody; Goat anti-mouse IgG (H&L), F(ab)'2 fragment - Affinity Pure, min x w/Human IgG or Serum Proteins, ALP Conjugate, Host: Goat, Format: Liq, Label: ALP, Applications: WB, ELISA
Catalog No. 50-271-791