Learn More
Immunoreagents Inc GOAT A-MOUSE IGG AP
Supplier:  Immunoreagents Inc GTXMU003J
Encompass_Preferred
Secondary Antibody; Goat anti-mouse IgG (H&L), F(ab)'2 fragment - Affinity Pure, min x w/Human IgG or Serum Proteins, Host: Goat, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC
Catalog No. 50-271-154