Learn More
Immunoreagents Inc GOAT A-MOUSE IGG BIO
Supplier:  Immunoreagents Inc GTXMU003E2BIO
Encompass_Preferred
Secondary Antibody; Goat anti-mouse IgG (H&L) - Affinity Pure, min x w/Bo, Eq, Hu, Pig or Rb Serum Proteins, Biotin Conjugate, Host: Goat, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-861