Learn More
Immunoreagents Inc GOAT A-MOUSE IGG DYLIGHT 550
Supplier:  Immunoreagents Inc GTXMU003J2550NHSX
Encompass_Preferred
Secondary Antibody; Goat anti-mouse IgG (H&L), F(ab)'2 fragment - Affinity Pure, min x w/Bo, Eq, Hu, Pig or Rb Serum Proteins, DyLight 550 Conjugate, Host: Goat, Format: Lyo, Label: DyL550, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-272-154