Learn More
Immunoreagents Inc GOAT A-MOUSE IGG HRP
Supplier:  Immunoreagents Inc GTXMU003L2HRPX
Encompass_Preferred
Secondary Antibody; Goat anti-mouse IgG (H&L), F(ab)'2 fragment, min x w/Bo, Gt, Hu, Rb or Rt IgG (highly absorbed against rat IgG), HRP Conjugate, Host: Goat, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-351