Learn More
Immunoreagents Inc GOAT A-MOUSE IGG HRP
Supplier:  Immunoreagents Inc GTXMU003GHRPX
Encompass_Preferred
Secondary Antibody; Goat anti-mouse IgG (H&L), F(ab)'2 fragment - Affinity Pure, HRP Conjugate, Host: Goat, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/IC
Catalog No. 50-271-576