Learn More
Immunoreagents Inc GOAT A-MOUSE IGG+IGM AP
Supplier:  Immunoreagents Inc GTXMU011E
Encompass_Preferred
Secondary Antibody; Goat anti-mouse IgG + IgM - Affinity Pure, min x w/Human IgG or Serum Proteins, Host: Goat, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC
Catalog No. 50-271-167