Learn More
Immunoreagents Inc GOAT A-MOUSE IGG+IGM FITC
Supplier:  Immunoreagents Inc GTXMU011DFITC
Encompass_Preferred
Secondary Antibody; Goat anti-mouse IgG + IgM - Affinity Pure, FITC Conjugate, Host: Goat, Format: Liq, Label: FITC, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-271-165