Learn More
Immunoreagents Inc GOAT A-MOUSE IGM MU BIO
Supplier:  Immunoreagents Inc GTXMU006DBIO
Encompass_Preferred
Secondary Antibody; Goat anti-mouse IgM (muchain) - Affinity Pure, Biotin Conjugate, Host: Goat, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-052