Learn More
Immunoreagents Inc GOAT A-PLASMINOGEN AP
Supplier:  Immunoreagents Inc GTXHU023D
Encompass_Preferred
Primary Antibodies: Goat anti-plasminogen - Affinity Pure, Host: Goat, Format: Liq, Label: None, Applications: WB, ELISA, IP, FC
Catalog No. 50-271-967