Learn More
Immunoreagents Inc GOAT A-RABBIT IGG ALP
Supplier:  Immunoreagents Inc GTXRB003E3ALP
Encompass_Preferred
Secondary Antibody; Goat anti-rabbit IgG (H&L) - Affinity Pure, min x w/Hu, Mu or Rt Serum Proteins, ALP Conjugate, Host: Goat, Format: Liq, Label: ALP, Applications: WB, ELISA
Catalog No. 50-271-803