Learn More
Immunoreagents Inc GOAT A-RABBIT IGG BIO
Supplier:  Immunoreagents Inc GTXRB004EBIO
Encompass_Preferred
Secondary Antibody; Goat anti-rabbit IgG Fc - Affinity Pure, min x/Bo, Eq or Hu Serum Proteins, Biotin Conjugate, Host: Goat, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-649