Learn More
Immunoreagents Inc GOAT A-RABBIT IGG BIO
Supplier:  Immunoreagents Inc GTXRB003EBIO
Encompass_Preferred
Secondary Antibody; Goat anti-rabbit IgG (H&L) - Affinity Pure, min x w/Bo, Hu or Mu IgG, Biotin Conjugate, Host: Goat, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-639