Learn More
Immunoreagents Inc GOAT A-RABBIT IGG DYLIGHT 550
Supplier:  Immunoreagents Inc GTXRB004D550NHSX
Encompass_Preferred
Secondary Antibody; Goat anti-rabbit IgG Fc - Affinity Pure, DyLight 550 Conjugate, Host: Goat, Format: Lyo, Label: DyL550, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-270-737