Learn More
Immunoreagents Inc GOAT A-RABBIT IGG DYLIGHT 633
Supplier:  Immunoreagents Inc GTXRB003E633NHSX
Encompass_Preferred
Secondary Antibody; Goat anti-rabbit IgG (H&L) - Affinity Pure, min x w/Bo, Hu or Mu IgG, DyLight 633 Conjugate, Host: Goat, Format: Lyo, Label: DyL633, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-270-718