Learn More
Immunoreagents Inc GOAT A-RABBIT IGG DYLIGHT 650
Supplier:  Immunoreagents Inc GTXRB003J650NHSX
Encompass_Preferred
Secondary Antibody; Goat anti-rabbit IgG (H&L), F(ab)'2 fragment - Affinity Pure, min x w/Bo, Hu or Mu IgG or Serum Proteins, DyLight 650 Conjugate, Host: Goat, Format: Lyo, Label: DyL650, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-272-062