Learn More
Immunoreagents Inc GOAT A-RABBIT IGG FITC
Supplier:  Immunoreagents Inc GTXRB003DFITC
Encompass_Preferred
Secondary Antibody; Goat anti-rabbit IgG (H&L) - Affinity Pure, FITC Conjugate, Host: Goat, Format: Liq, Label: FITC, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-272-175