Learn More
Immunoreagents Inc GOAT A-RABBIT IGG HRP
Supplier:  Immunoreagents Inc GTXRB004EHRPX
Encompass_Preferred
Secondary Antibody; Goat anti-rabbit IgG Fc - Affinity Pure, min x/Bo, Eq or Hu Serum Proteins, HRP Conjugate, Host: Goat, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-272-167