Learn More
Immunoreagents Inc GOAT A-RABBIT IGG HRP
Supplier:  Immunoreagents Inc GTXRB003E2HRPX
Encompass_Preferred
Secondary Antibody; Goat anti-rabbit IgG (H&L) - Affinity Pure, min x w/Human Serum Proteins, HRP Conjugate, Host: Goat, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-272-164