Learn More
Immunoreagents Inc GOAT A-RAT IGG ALP
Supplier:  Immunoreagents Inc GTXRT003E2ALP
Encompass_Preferred
Secondary Antibody; Goat anti-rat IgG (H&L) - Affinity Pure, min x w/Hu or Mu IgG (highly absorbed against Mu IgG), ALP Conjugate, Host: Goat, Format: Liq, Label: ALP, Applications: WB, ELISA
Catalog No. 50-271-217