Learn More
Immunoreagents Inc GOAT A-RAT IGG AP
Supplier:  Immunoreagents Inc GTXRT003F
Encompass_Preferred
Secondary Antibody; Goat anti-rat IgG (H&L) - Affinity Pure, min x w/Bo, Gt, Hu Rb or Mu IgG, Host: Goat, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC
Catalog No. 50-270-870