Learn More
Immunoreagents Inc GOAT A-RAT IGG AP
Supplier:  Immunoreagents Inc GTXRT003L2
Encompass_Preferred
Secondary Antibody: Goat anti-rat IgG (H&L), F(ab)'2 fragment - Affinity Pure, min x w/Hu, Mu IgG (highly absorbed against Mu IgG), Host: Goat, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC
Catalog No. 50-271-354