Learn More
Immunoreagents Inc GOAT A-RAT IGG BIO
Supplier:  Immunoreagents Inc GTXRT003L2BIO
Encompass_Preferred
Secondary Antibody: Goat anti-rat IgG (H&L), F(ab)'2 fragment - Affinity Pure, min x w/Hu, Mu IgG (highly absorbed against Mu IgG), Biotin Conjugate, Host: Goat, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-690