Learn More
Immunoreagents Inc GOAT A-RAT IGG BIO
Supplier:  Immunoreagents Inc GTXRT003EBIO
Encompass_Preferred
Secondary Antibody; Goat anti-rat IgG (H&L) - Affinity Pure, min x w/Hu or Mu IgG, Biotin Conjugate, Host: Goat, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-272-031