Learn More
Biomatik Corporation Goat Adrenocorticotropic Hormone (ACTH) ELISA Kit
Supplier:  Biomatik Corporation EKU02176
Encompass_Preferred
Goat Adrenocorticotropic Hormone (ACTH) ELISA Kit for detecting ACTH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Goat (Capra aegagrus hircus). Detection Range: 12.35-1000pg/ml. Sensitivity: 4.92pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: A0A452FYE1. Gene ID: 102182514. Target Synonyms: Corticotropin; Adrenocorticotropin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7012