Learn More
American Research Products Inc Goat Albumin Alb ELISA Kit
Catalog No. 5000328629
Encompass
Goat Albumin (Alb) ELISA Kit from CUSABIO Detection Range 0 17 g/mL-350 g/mL Sensitivity 0 17 g/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28629 Supplier American Research Products Inc Supplier No. CSBE15758G
Goat Albumin (Alb) ELISA Kit from CUSABIO Detection Range 0 17 g/mL-350 g/mL Sensitivity 0 17 g/mL Sample serum plasma tissue homogenates Method Competitive