Learn More
Innovative Research Inc Goat Albumin Ply Aff Pur 1mg
Catalog No. 502744374
Encompass_Preferred
Goat Anti Human Albumin Polyclonal Affinity Purified 1mg - Goat Anti Human Albumin Polyclonal Affinity Purified from Innovative Research is a polyclonal antibody in Liquid format, buffered in Phospate buffered saline, pH 7.2 with 0.1% sodium azide. Goats were immunized with highly purified human serum albumin in Freund™s adjuvant. Anti-Albumin antibodies were immunoaffinity purified off a human albumin affinity immunoabsorbant. More Details Host Goat Species Human Target anti-Human Albumin Purification Affinity Purified Clonality Polyclonal Storage Conditions 4C This product can be used for a wide range of research applications, including ELISA Assay Dependent, IP 2 ug/mg, LFD - Capture, WB 0.5 - 0.05 ug/ml.
Catalog No. 50-274-4374 Supplier Innovative Research Inc Supplier No. IGTAHUALBAP1MG
Goat Anti Human Albumin Polyclonal Affinity Purified 1mg - Goat Anti Human Albumin Polyclonal Affinity Purified from Innovative Research is a polyclonal antibody in Liquid format, buffered in Phospate buffered saline, pH 7.2 with 0.1% sodium azide. Goats were immunized with highly purified human serum albumin in Freund™s adjuvant. Anti-Albumin antibodies were immunoaffinity purified off a human albumin affinity immunoabsorbant. More Details Host Goat Species Human Target anti-Human Albumin Purification Affinity Purified Clonality Polyclonal Storage Conditions 4C This product can be used for a wide range of research applications, including ELISA Assay Dependent, IP 2 ug/mg, LFD - Capture, WB 0.5 - 0.05 ug/ml.