Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anit-Rabbit IgG, Whole Molecule Polyclonal Secondary Antibody, HRP, MP Biomedicals

Catalog No. ICN55689 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN55689 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN55689 Supplier MP Biomedicals, Inc Supplier No. 0855689
Add to Cart
Add to Cart

Horseradish peroxidase (HRP)-conjugated goat affinity purified antibody to rabbit IgG (whole molecule) and buffer salts.

Host Species: Goat
Species Reactivity: Rabbit

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG, Whole Molecule
Classification Polyclonal
Conjugate HRP
Host Species Goat
Quantity 2 mL
Primary or Secondary Secondary
Target Species Rabbit
Form Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.