Learn More
Immunoreagents Inc GOAT ANTI-BIOTIN ALKPHOS 1MG
Supplier:  Immunoreagents Inc GTXOT070DALP
Encompass_Preferred
Primary Antibody; Goat anti-biotin, Affinity Pure, ALP Conjugate, Host: Goat, Format: Liq, Label: ALP, Applications: ELISA, WB
Catalog No. 50-271-764