Learn More
Goat anti-Bovine IgG whole molecule, Polyclonal, Secondary Antibody, Abnova™
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Antigen | Goat anti-Bovine IgG whole molecule |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Goat anti-bovine IgG recognizes bovine IgG whole molecule. This secondary antibody was purified using delipidation and defibrination. |
| Dilution | The optimal working dilution should be determined by the end user. |
| Formulation | Lyophilized from 10mM KH2PO4, 150mM NaCl, pH 7.2 |
| Host Species | Goat |
| Immunogen | Bovine IgG whole molecule |
| Purification Method | This product was prepared from monospecific antiserum by a delipidation and defibrination. Assay by immunoelectrophoresis resulted in a single precipitin arc against bovine IgG and bovine serum. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.