Learn More
Vector Laboratories Goat Anti-Cat IgG Antibody (H+L), Biotinylated, 1.5mg
SDP
Supplier:  Vector Laboratories BA90001.5
Encompass
Biotinylated Goat Anti-Cat IgG Antibody is used in an avidin-biotin or streptavidin-biotin detection system. These antibodies can be used for tissue and cell staining, ELISAs, and blots. Features: Affinity-purified, ultrapure, high affinity antibody Thoroughly adsorbed against serum and immunoglobulins from potentially interfering species . Recognizes both heavy and light chains (H+L) Biotinylated to ensure the maximum degree of labeling without compromising the specificity or affinity of the antibody.
Catalog No. 50-204-5622