Learn More
Goat anti-Chicken IgG whole molecule, Texas Red, Polyclonal, Secondary Antibody, Abnova™
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Antigen | Goat anti-Chicken IgG whole molecule |
| Applications | Flow Cytometry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Texas Red |
| Description | Goat anti-chicken IgG recognizes chicken IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Texas Red. |
| Dilution | The optimal working dilution should be determined by the end user. |
| Formulation | Lyophilized |
| Host Species | Goat |
| Immunogen | Chicken IgG whole molecule |
| Purification Method | This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.