Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat Anti-Chicken IgY (Fc) Antibody (FITC), ProSci
SDP

Catalog No. 16823377 Shop All ProSci Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-823-377 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-823-377 Supplier ProSci LLC Supplier No. XW91020100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Control Antibody

Goat Anti-Chicken IgY (Fc) Antibody specifically detects Goat Anti-Chicken IgY (Fc) in Chicken samples. It is validated for ELISA, Western Blot

Specifications

Antigen Goat Anti-Chicken IgY (Fc)
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Fluorescein
Dilution The affinity-purified antibodies react only with chicken IgY and not IgM or IgA immunoglobulins by immunodiffusion and IEP techniques. Suggested immunostaining working dilution for this lot should start from 1:80 or up when used as 2nd antibody. Optimal conjugate working dilutions should be determined for your particular assay conditions.
Formulation PBS, pH 7.2 with 0.2% BSA
Host Species Goat
Purification Method Affinity
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Chicken
Content And Storage 4 °C, avoid frequent freeze-thaw
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.