Learn More
Invitrogen™ Goat anti-Chicken IgY (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ Plus 647
Shop All Thermo Scientific ProductsDescription
To minimize cross-reactivity, the goat anti-chicken IgY whole antibodies have been affinity-purified and cross-adsorbed against chicken serum containing non-immunoglobulin chicken serum proteins. Cross-adsorption or pre-adsorption is a purification step to increase specificity of the antibody resulting in higher sensitivity and less background staining. The secondary antibody solution is passed through a column matrix containing immobilized serum proteins from potentially cross-reactive species. Only the nonspecific-binding secondary antibodies are captured in the column, and the highly specific secondaries flow through. The benefits of this extra step are apparent in multiplexing/multicolor-staining experiments (e.g., flow cytometry) where there is potential cross-reactivity with other primary antibodies or in tissue/cell fluorescent staining experiments where there may be the presence of endogenous immunoglobulins. Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Because staining protocols vary with application, the appropriate dilution of antibody should be determined empirically. Specificity: This antibody binds to heavy chains on chicken IgY and light chains on all chicken immunoglobulins. This antibody does not bind non-immuno...
Specifications
Specifications
| Antigen | Chicken IgY (H+L) Cross-Adsorbed |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Alexa Fluor Plus 647 |
| Formulation | Proprietary buffer with 0.016% Bromonitrodioxane, 0.016% Methylisothiazolone; pH 6.5 |
| Host Species | Goat |
| Immunogen | Purified Chicken IgY, Heavy and Light chains. |
| Purification Method | Affinity chromatography |
| Quantity | 1 mg |
| Show More |
Frequently Asked Questions (FAQs)
The TAC currently does not provide pathway analysis for small panels such as the Oncomine Immune Response Research Assay.
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.