Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat Anti-Chicken IgY NorthernLights™ NL493-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. NL018
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NL018 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NL018 Supplier R&D Systems Supplier No. NL018
This item is not returnable. View return policy

Goat Polyclonal Antibody

IgY Polyclonal specifically detects IgY in Chicken samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry.

Specifications

Antigen IgY
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Conjugate NorthernLights 493 (NL493)
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry
Host Species Goat
Purification Method Affinity Purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity Detects chicken IgY heavy and light chains. In direct ELISA, less than 1% cross-reactivity with human IgG, rabbit IgG, rat IgG, donkey IgG and mouse IgG is observed.
Target Species Chicken
Content And Storage Store the unopened product at 2 - 8degrees C. Do not use past expiration date. Protect from light.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.