Learn More
Immunology Consultants Affinity Purified Goat anti-CHO Phospholipase B-like 2 protein (PLBL2) Antibody, 0.5 mg
SDP
Supplier:  Immunology Consultants GPLB65ALY
Encompass_Preferred
Goats were immunized with CHO Phospholipase B-Like 2 (PLBL2) of recombinant origin and antiserum were collected. The antibody was immunoaffinity purified off an antigen containing immunosorbent. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with CHO Phospholipase B-Like 2 (PLBL2) of recombinant origin and PLBL2 secreted by a null CHO cell line as determined by ELISA and western blot. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-224-8915