Learn More
List Biological Laboratories Inc Anti-Cholera Toxin B Subunit (Goat), 0.1 mL
SDP
Catalog No. NC9925410
Encompass_Preferred
Since 1978, List Labs products are unsurpassed in quality, consistency, and reliability for the success of your research and development project! Cholera toxin is an oligomeric protein of MW 84,000 daltons which consists of a single A subunit surrounded by five B subunits. It is a potent activator of adenylate cyclase and is the pathogenic agent responsible for the symptoms of cholera. Cholera toxin has become a powerful research tool not only in microbiology, but in the fields of physiology, cell biology and biochemistry, as well. Because of the effect on adenylate cyclase, cholera toxin is frequently used for the study of signal transduction mechanisms. In addition, cholera toxin acts as an adjuvant through the stimulation of B-lymphocytes. High titer polyclonal chloragenoid from goat, suitable for use in either toxin neutralization or binding assays is provided as a lyophilized powder containing 0.1% NaN2 as a preservative.
Catalog No. NC9925410 Supplier List Biological Laboratories Inc Supplier No. 703
Since 1978, List Labs products are unsurpassed in quality, consistency, and reliability for the success of your research and development project! Cholera toxin is an oligomeric protein of MW 84,000 daltons which consists of a single A subunit surrounded by five B subunits. It is a potent activator of adenylate cyclase and is the pathogenic agent responsible for the symptoms of cholera. Cholera toxin has become a powerful research tool not only in microbiology, but in the fields of physiology, cell biology and biochemistry, as well. Because of the effect on adenylate cyclase, cholera toxin is frequently used for the study of signal transduction mechanisms. In addition, cholera toxin acts as an adjuvant through the stimulation of B-lymphocytes. High titer polyclonal chloragenoid from goat, suitable for use in either toxin neutralization or binding assays is provided as a lyophilized powder containing 0.1% NaN2 as a preservative.